Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010484.2 | NP_034614.2 | 13, 14 | 2011 | ||
GenBank mRNA | AF013604.1 | - | 12, 13 | 1824 | ||
AK008410.1 | - | 2, 3 | 211 | |||
AY765218.1 | - | 2, 3 | 269 | |||
BC108978.1 | - | 13, 14 | 1993 | 40047534 | ||
BC108979.1 | - | 13, 14 | 1993 | 40047537 | ||
X66119.1 | - | 12, 13 | 1605 |