Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010510.1 | NP_034640.1 | 1 | 468 | ||
GenBank mRNA | BC119395.1 | - | 1 | 468 | 8734144 | |
BC119397.1 | - | 1 | 468 | 8734146 | ||
K00020.1 | - | 1 | 468 | |||
M27321.1 | - | 57 |