Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010551.3 | NP_034681.2 | 19 | 4960 | ||
GenBank mRNA | AF017111.1 | - | 7 | 2843 | ||
AF175292.1 | - | 19 | 4961 | |||
AK038014.1 | - | 1802 | ||||
AK038319.1 | - | 7 | 2876 | |||
AK134007.1 | - | 7 | 2792 | |||
AK169551.1 | - | 7 | 2863 | |||
AK220455.1 | - | 19 | 4971 | |||
BC058709.1 | - | 19 | 4993 | 6841305 |