Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052997.2 | NP_443723.2 | 2, 3 | 275 | ||
XM_011519757.2 | XP_011518059.1 | 2, 3 | 378 | |||
XM_011519758.2 | XP_011518060.1 | 2, 3 | 378 | |||
GenBank mRNA | AF269087.1 | - | 2, 3 | 275 | ||
DN831985.1 | - | 1, 2 | 65 |