Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010576.3 | NP_034706.3 | 8, 9 | 1138 | ||
GenBank mRNA | AB850880.1 | - | 8, 9 | 903 | ||
AF109136.1 | - | 8, 9 | 974 | |||
AK051676.1 | - | 8, 9 | 1064 | |||
AK078842.1 | - | 8, 9 | 843 | |||
AK089898.1 | - | 7, 8 | 938 | |||
AK156015.1 | - | 8, 9 | 1034 | |||
AK156517.1 | - | 8, 9 | 908 | |||
BC068313.1 | - | 8, 9 | 918 | 30093248 | ||
U97151.1 | - | 6, 7 | 506 | |||
X53176.1 | - | 8, 9 | 1061 |