Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281965.1 | NP_001268894.1 | 2 | 269 | ||
NM_001281966.1 | NP_001268895.1 | 2 | 269 | |||
NM_001281967.1 | NP_001268896.1 | 2 | 269 | |||
NM_001281968.1 | NP_001268897.1 | 2 | 266 | |||
NM_010583.3 | NP_034713.2 | 2 | 269 | |||
GenBank mRNA | AK020772.1 | - | 2 | 217 | ||
AK079693.1 | - | 2 | 217 | |||
AK083380.1 | - | 2 | 239 | |||
AK153613.1 | - | 2 | 216 | |||
AK153692.1 | - | 2 | 216 | |||
BB843988.1 | - | 2 | 269 | |||
BC028478.1 | - | 2 | 211 | |||
BC128374.1 | - | 2 | 195 | 40111687 | ||
BC128375.1 | - | 2 | 192 | 40111688 | ||
D14042.1 | - | 2 | 209 | |||
L00619.1 | - | 2 | 256 | |||
L05631.1 | - | 2 | 235 | |||
L10628.1 | - | 2 | 237 |