Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010612.2 | NP_034742.2 | 3 | 481 | ||
GenBank mRNA | AK054510.1 | - | 3 | 514 | ||
AK141938.1 | - | 3 | 504 | |||
BC020530.1 | - | 3 | 481 | 4238984 | ||
EU884114.1 | - | 3 | 511 | |||
S53103.1 | - | 3 | 492 | |||
X59397.1 | - | 3 | 428 | |||
X70842.1 | - | 3 | 506 |