Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190978.1 | NP_001177907.1 | 2 | 540 | ||
NM_010623.4 | NP_034753.1 | 2 | 540 | |||
GenBank mRNA | AB001424.1 | - | 1 | 30 | ||
AB008867.1 | - | 2 | 538 | |||
AK046890.1 | - | 2 | 293 | |||
AK077868.1 | - | 2 | 321 |