Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010637.3 | NP_034767.2 | 3 | 750 | ||
GenBank mRNA | AA389216.1 | - | 3 | 282 | ||
AK144942.1 | - | 3 | 748 | |||
AK155343.1 | - | 3 | 748 | |||
BC010301.1 | - | 3 | 612 | 3156339 | ||
CF163360.1 | - | 2 | 94 | |||
JF277566.1 | - | 2 | 146 | |||
U20344.1 | - | 3 | 456 | |||
U70662.1 | - | 3 | 744 |