Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252577.1 | NP_001239506.1 | 2 | 231 | ||
NM_010649.3 | NP_034779.1 | 2 | 243 | |||
GenBank mRNA | AK080171.1 | - | 2 | 232 | ||
AK145141.1 | - | 1 | 102 | |||
AY860975.1 | - | 1 | 14 | |||
L78247.1 | - | 1 | 86 | |||
U10090.1 | - | 1 | 153 |