Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010655.3 | NP_034785.1 | 2, 3 | 264 | ||
GenBank mRNA | AI664586.1 | - | 2, 3 | 264 | ||
AK075952.1 | - | 2, 3 | 203 | |||
AK088496.1 | - | 2, 3 | 203 | |||
AK139850.1 | - | 2, 3 | 201 | |||
AK147135.1 | - | 2, 3 | 203 | |||
AK165577.1 | - | 2, 3 | 205 | |||
AK166577.1 | - | 2, 3 | 207 | |||
AK166790.1 | - | 2, 3 | 203 | |||
AK166982.1 | - | 2, 3 | 201 | |||
AK167551.1 | - | 2, 3 | 203 | |||
AK169283.1 | - | 2, 3 | 203 | |||
BC003274.1 | - | 2, 3 | 186 | 3499880 | ||
BC006720.1 | - | 2, 3 | 184 | 3711035 | ||
BC082280.1 | - | 2, 3 | 158 | 6334979 | ||
BC086789.1 | - | 2, 3 | 182 | |||
BC094011.1 | - | 2, 3 | 176 | 5035738 | ||
BC106110.1 | - | 2, 3 | 177 | |||
D55720.1 | - | 2, 3 | 251 | |||
U12270.1 | - | 2, 3 | 189 | |||
U34229.1 | - | 1, 2 | 74 |