Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010689.2 | NP_034819.1 | 6, 7 | 647 | ||
GenBank mRNA | AF036907.1 | - | 6, 7 | 647 | ||
AK088763.1 | - | 6, 7 | 556 | |||
AK134003.1 | - | 6, 7 | 599 | |||
BC013337.1 | - | 6, 7 | 624 | 4191983 | ||
BC052340.1 | - | 6, 7 | 592 | 30077071 |