Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010575.2 | NP_034705.2 | 2, 3 | 490 | ||
GenBank mRNA | AF166384.1 | - | 2, 3 | 368 | ||
AF170316.1 | - | 2, 3 | 351 | |||
AK142289.1 | - | 2, 3 | 352 | |||
AK154619.1 | - | 2, 3 | 491 | |||
AK156872.1 | - | 2, 3 | 353 | |||
AK157012.1 | - | 2, 3 | 352 | |||
AK158443.1 | - | 2, 3 | 337 | |||
BC120493.1 | - | 2, 3 | 340 | 40129416 | ||
BC137570.1 | - | 2, 3 | 340 | 8860589 |