Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010576.3 | NP_034706.3 | 9 | 1244 | ||
GenBank mRNA | AB850880.1 | - | 9 | 1009 | ||
AF109136.1 | - | 9 | 1080 | |||
AK051676.1 | - | 9 | 1170 | |||
AK089898.1 | - | 8 | 1044 | |||
AK156015.1 | - | 9 | 1140 | |||
AK156517.1 | - | 9 | 1014 | |||
BC068313.1 | - | 9 | 1024 | 30093248 | ||
X53176.1 | - | 9 | 1167 |