Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010577.3 | NP_034707.3 | 2 | 516 | ||
GenBank mRNA | AK142241.1 | - | 2 | 359 | ||
AK144921.1 | - | 2 | 302 | |||
AK148138.1 | - | 2 | 356 | |||
AK152594.1 | - | 2 | 314 | |||
BC050943.1 | - | 2 | 532 | 6821577 | ||
BC053431.1 | - | 3 | 577 | |||
BC059829.1 | - | 2 | 532 | 6821577 | ||
X79003.1 | - | 2 | 473 |