Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038607.2 | NP_001033696.1 | 3 | 485 | ||
NM_010600.3 | NP_034730.1 | 3 | 485 | |||
GenBank mRNA | AK140193.1 | - | 3 | 391 | ||
AK147461.1 | - | 3 | 479 | |||
BC109012.1 | - | 3 | 344 | 40048708 | ||
BC109013.1 | - | 3 | 344 | 40048709 | ||
CJ104826.1 | - | 3 | 487 | |||
U04294.1 | - | 3 | 367 |