Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025584.2 | NP_001020755.1 | 3 | 694 | ||
NM_001025585.2 | NP_001020756.1 | 3 | 694 | |||
NM_001025590.1 | NP_001020761.1 | 2 | 370 | |||
NM_010606.2 | NP_034736.2 | 3 | 694 | |||
GenBank mRNA | AB029502.1 | - | 3 | 456 | ||
AK015481.1 | - | 2 | 370 | |||
AK044218.1 | - | 4 | 809 | |||
AK081109.1 | - | 3 | 429 | |||
AK139056.1 | - | 3 | 698 | |||
BC120794.1 | - | 3 | 604 | 40129717 | ||
BC125565.1 | - | 3 | 604 | 40130080 | ||
D86040.1 | - | 3 | 372 | |||
U11859.1 | - | 3 | 653 | |||
U37253.1 | - | 3 | 653 | |||
U51122.1 | - | 3 | 646 | |||
U51123.1 | - | 3 | 646 | |||
U51124.1 | - | 3 | 646 | |||
U51125.1 | - | 3 | 581 | |||
U51126.1 | - | 2 | 358 |