Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145831.1 | NP_001139303.1 | 8 | 983 | ||
NM_001145832.1 | NP_001139304.1 | 4 | 467 | |||
NM_010631.2 | NP_034761.3 | 6 | 810 | |||
GenBank mRNA | AF013118.2 | - | 2 | 446 | ||
AK132895.1 | - | 4 | 467 | |||
AK166482.1 | - | 8 | 963 | |||
BC016118.1 | - | 1 | 14 | |||
BC070429.1 | - | 6 | 810 | 6810893 | ||
BI737872.1 | - | 4 | 402 | |||
D49546.1 | - | 6 | 825 |