Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010637.3 | NP_034767.2 | 4 | 1828 | ||
GenBank mRNA | AK141244.1 | - | 2 | 740 | ||
AK144942.1 | - | 4 | 1826 | |||
AK155343.1 | - | 4 | 1826 | |||
BC010301.1 | - | 4 | 1690 | 3156339 | ||
JF277566.1 | - | 3 | 1224 | |||
U20344.1 | - | 4 | 1534 | |||
U70662.1 | - | 4 | 1822 |