Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010655.3 | NP_034785.1 | 3 | 393 | ||
GenBank mRNA | AI664586.1 | - | 3 | 393 | ||
AK075952.1 | - | 3 | 332 | |||
AK088496.1 | - | 3 | 332 | |||
AK139850.1 | - | 3 | 330 | |||
AK147135.1 | - | 3 | 332 | |||
AK165577.1 | - | 3 | 334 | |||
AK166577.1 | - | 3 | 336 | |||
AK166790.1 | - | 3 | 332 | |||
AK166982.1 | - | 3 | 330 | |||
AK167551.1 | - | 3 | 332 | |||
AK169283.1 | - | 3 | 332 | |||
BC003274.1 | - | 3 | 315 | 3499880 | ||
BC006720.1 | - | 3 | 313 | 3711035 | ||
BC082280.1 | - | 3 | 287 | 6334979 | ||
BC086789.1 | - | 3 | 311 | |||
BC094011.1 | - | 3 | 305 | 5035738 | ||
BC106110.1 | - | 3 | 305 | |||
D55720.1 | - | 3 | 380 | |||
U12270.1 | - | 3 | 318 | |||
U34229.1 | - | 2 | 203 |