Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010663.2 | NP_034793.1 | 3, 4 | 701 | ||
GenBank mRNA | AB013608.1 | - | 3, 4 | 702 | ||
BC032161.1 | - | 3, 4 | 748 | |||
BC132454.1 | - | 3, 4 | 736 | 40130731 | ||
BC132456.1 | - | 3, 4 | 736 | 40130733 |