Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278622.1 | NP_001265551.1 | 5, 6 | 896 | ||
NM_002873.1 | NP_002864.1 | 4, 5 | 1001 | |||
NM_133338.2 | NP_579916.1 | 6, 7 | 897 | |||
NM_133339.2 | NP_579917.1 | 3, 4 | 454 | |||
NM_133340.2 | NP_579918.1 | 3, 4 | 487 | |||
NM_133341.2 | NP_579919.1 | 3, 4 | 413 | |||
NM_133342.2 | NP_579920.1 | 5, 6 | 874 | |||
NM_133343.1 | NP_579921.1 | 5, 6 | 839 | |||
NM_133344.2 | NP_579922.1 | 4, 5 | 656 | |||
XM_017009679.1 | XP_016865168.1 | 5, 6 | 716 | |||
XM_017009680.1 | XP_016865169.1 | 4, 5 | 770 | |||
XM_017009681.1 | XP_016865170.1 | 4, 5 | 628 | |||
GenBank mRNA | AF017748.1 | - | 4, 5 | 626 | ||
AF076838.1 | - | 4, 5 | 1001 | |||
AF085736.2 | - | 3, 4 | 454 | |||
AF098533.1 | - | 3, 4 | 433 | |||
AF098534.1 | - | 3, 4 | 359 | |||
AF112263.1 | - | 3, 4 | 478 | |||
AF126424.1 | - | 6, 7 | 896 | |||
AJ001642.1 | - | 4, 5 | 665 | |||
AJ004977.1 | - | 4, 5 | 661 | |||
AK292487.1 | - | 5, 6 | 874 | |||
AL122068.1 | - | 5, 6 | 839 | |||
BC032304.1 | - | 3, 4 | 467 | 5170876 | ||
BX537441.1 | - | 5, 6 | 896 |