Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136069.2 | NP_001129541.2 | 6 | 1023 | ||
NM_010699.2 | NP_034829.1 | 6 | 830 | |||
GenBank mRNA | AK089929.1 | - | 6 | 794 | ||
AK146121.1 | - | 6 | 791 | |||
AK146980.1 | - | 2 | 205 | |||
AK149919.1 | - | 6 | 791 | |||
AK150156.1 | - | 6 | 794 | |||
AK167789.1 | - | 6 | 792 | |||
AK167944.1 | - | 6 | 792 | |||
AK168346.1 | - | 6 | 791 | |||
AK168920.1 | - | 6 | 796 | |||
AK170386.1 | - | 6 | 795 | |||
AK170708.1 | - | 6 | 792 | |||
AK189424.1 | - | 1 | 73 | |||
AK194735.1 | - | 2 | 188 | |||
AK198750.1 | - | 1 | 72 | |||
AK200149.1 | - | 2 | 139 | |||
AK203954.1 | - | 1 | 72 | |||
AK209400.1 | - | 2 | 261 | |||
AK216074.1 | - | 1 | 72 | |||
AK217915.1 | - | 1 | 72 | |||
AK220005.1 | - | 1 | 73 | |||
BC005509.1 | - | 5 | 642 | |||
BC066858.1 | - | 5 | 448 | |||
BC094019.1 | - | 6 | 747 | 4195220 | ||
BC094428.1 | - | 6 | 784 | 30700835 | ||
M17516.1 | - | 1 | 89 | |||
U13687.1 | - | 6 | 890 |