Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010577.3 | NP_034707.3 | 26, 27 | 2955 | ||
GenBank mRNA | AK142241.1 | - | 26, 27 | 2798 | ||
AK144921.1 | - | 26, 27 | 2742 | |||
AK148138.1 | - | 26, 27 | 2796 | |||
BC050943.1 | - | 26, 27 | 2971 | 6821577 | ||
BC053431.1 | - | 27, 28 | 3004 | |||
BC059829.1 | - | 26, 27 | 2971 | 6821577 | ||
X15203.1 | - | 8, 9 | 801 | |||
X79003.1 | - | 26, 27 | 2912 |