Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190978.1 | NP_001177907.1 | 5 | 1037 | ||
NM_010623.4 | NP_034753.1 | 5 | 1037 | |||
GenBank mRNA | AB008867.1 | - | 5 | 1035 | ||
AK046890.1 | - | 5 | 790 | |||
AK077868.1 | - | 5 | 818 |