Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145831.1 | NP_001139303.1 | 22 | 3187 | ||
NM_001145832.1 | NP_001139304.1 | 18 | 2701 | |||
NM_010631.2 | NP_034761.3 | 20 | 3044 | |||
GenBank mRNA | AF013118.2 | - | 16 | 2677 | ||
AK135926.1 | - | 15 | 2243 | |||
AK166482.1 | - | 22 | 3149 | |||
BC004069.1 | - | 12 | 1774 | |||
BC016118.1 | - | 15 | 2248 | |||
BC023374.1 | - | 13 | 1836 | |||
BC070429.1 | - | 20 | 3044 | 6810893 |