Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010637.3 | NP_034767.2 | 5 | 2590 | ||
GenBank mRNA | AK155343.1 | - | 5 | 2584 | ||
BC091737.1 | - | 1 | 585 | |||
U20344.1 | - | 5 | 2293 | |||
U70662.1 | - | 5 | 2582 |