Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010656.2 | NP_034786.1 | 3 | 1383 | ||
GenBank mRNA | AK148179.1 | - | 3 | 1385 | ||
AK162864.1 | - | 3 | 1408 | |||
AK164529.1 | - | 3 | 1355 | |||
BC021484.1 | - | 3 | 1411 | 5344514 | ||
U02487.1 | - | 3 | 1383 |