Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010662.1 | NP_034792.1 | 7, 8 | 1251 | ||
GenBank mRNA | AK004635.1 | - | 7, 8 | 1322 | ||
AK009677.1 | - | 7, 8 | 812 | |||
AK010156.1 | - | 7, 8 | 1322 | |||
AK132640.1 | - | 7, 8 | 1323 | |||
X03492.1 | - | 7, 8 | 876 |