Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010696.3 | NP_034826.2 | 21 | 2248 | ||
GenBank mRNA | AK036727.1 | - | 21 | 2306 | ||
AK168134.1 | - | 21 | 2264 | |||
AK170491.1 | - | 21 | 2525 | |||
BC006948.1 | - | 21 | 2250 | 3603181 |