Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010746.3 | NP_034876.2 | 3 | 223 | ||
GenBank mRNA | AJ223765.1 | - | 3 | 212 | ||
AK079401.1 | - | 3 | 224 | |||
AK172251.1 | - | 2 | 256 | |||
BC042788.1 | - | 3 | 193 | |||
BC115364.2 | - | 3 | 224 | 40045898 |