Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010749.6 | NP_034879.2 | 7 | 1246 | ||
GenBank mRNA | AK004656.1 | - | 6 | 2889 | ||
AK133943.1 | - | 7 | 1252 | |||
AK145913.1 | - | 7 | 1258 | |||
AK149177.1 | - | 7 | 1276 | |||
AK151607.1 | - | 7 | 1307 | |||
AK153140.1 | - | 7 | 1257 | |||
AK153302.1 | - | 7 | 1327 | |||
AK153474.1 | - | 7 | 1267 | |||
AK190285.1 | - | 1 | 183 | |||
AK195131.1 | - | 1 | 138 | |||
AK215662.1 | - | 1 | 256 | |||
BC027210.1 | - | 7 | 1201 | |||
BC046956.1 | - | 7 | 1246 | |||
BC080811.1 | - | 7 | 1246 | 6438831 | ||
M63286.1 | - | 7 | 1154 | |||
X56831.1 | - | 7 | 1187 | |||
X64068.1 | - | 7 | 1249 |