Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010752.3 | NP_034882.2 | 3 | 223 | ||
GenBank mRNA | AF083812.1 | - | 3 | 230 | ||
AF083813.1 | - | 3 | 230 | |||
AK136981.1 | - | 3 | 225 | |||
BC010702.1 | - | 3 | 221 | 3156639 | ||
BI735882.1 | - | 3 | 222 |