Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010758.2 | NP_034888.1 | 4 | 495 | ||
GenBank mRNA | AK045564.1 | - | 3 | 605 | ||
BC101944.1 | - | 4 | 468 | 40039200 | ||
BC103521.1 | - | 4 | 468 | 40039204 | ||
BC103522.1 | - | 4 | 468 | |||
BC103523.1 | - | 4 | 468 | |||
M31811.1 | - | 4 | 495 |