Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282737.1 | NP_001269666.1 | 2, 3 | 251 | ||
GenBank mRNA | AF007862.1 | - | 2, 3 | 238 | ||
AF035939.1 | - | 2, 3 | 198 | |||
AK011512.1 | - | 2, 3 | 190 | |||
BC018176.1 | - | 2, 3 | 189 | 3587774 | ||
BY134232.1 | - | 2, 3 | 252 |