Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285853.1 | NP_001272782.1 | 3 | 425 | ||
NM_001285854.1 | NP_001272783.1 | 3 | 422 | |||
NM_001285855.1 | NP_001272784.1 | 3 | 490 | |||
NM_010768.2 | NP_034898.1 | 4 | 558 | |||
GenBank mRNA | AK013606.1 | - | 3 | 239 | ||
AK018182.1 | - | 3 | 277 | |||
AK079816.1 | - | 3 | 218 | |||
AK158883.1 | - | 3 | 283 | |||
BC060086.1 | - | 3 | 379 | |||
BC111864.1 | - | 3 | 236 | 40046807 | ||
D45243.1 | - | 3 | 490 | |||
L27738.1 | - | 4 | 312 | |||
S77473.1 | - | 3 | 237 | |||
U05210.1 | - | 3 | 211 |