Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288586.1 | NP_001275515.1 | 4 | 411 | ||
NM_010786.4 | NP_034916.1 | 5 | 496 | |||
GenBank mRNA | AK004719.1 | - | 5 | 455 | ||
AK004881.1 | - | 5 | 423 | |||
AK088638.1 | - | 5 | 426 | |||
AK150530.1 | - | 4 | 284 | |||
AK152685.1 | - | 4 | 284 | |||
AK160069.1 | - | 4 | 287 | |||
AK168690.1 | - | 5 | 496 | |||
BC050902.1 | - | 5 | 412 | 6415937 | ||
BC092270.1 | - | 5 | 449 | 4018419 | ||
CN703560.1 | - | 4 | 353 | |||
U47934.1 | - | 3 | 202 | |||
X58876.1 | - | 5 | 403 |