Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193271.1 | NP_001180200.1 | 5 | 1203 | ||
NM_010789.3 | NP_034919.1 | 5 | 1203 | |||
GenBank mRNA | AB221660.1 | - | 4 | 466 | ||
AK032947.1 | - | 4 | 1014 | |||
AK049720.1 | - | 4 | 1015 | |||
AK132298.1 | - | 5 | 983 | |||
AK134321.1 | - | 5 | 1198 | |||
AK140748.1 | - | 5 | 1207 | |||
AK168684.1 | - | 5 | 1195 | |||
BC023689.1 | - | 5 | 1199 | 5344307 | ||
U33629.1 | - | 5 | 914 | |||
U33630.1 | - | 5 | 914 |