Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 3 | 274 | ||
GenBank mRNA | AK010498.1 | - | 3 | 241 | ||
AK011932.1 | - | 3 | 274 | |||
AK129076.1 | - | 3 | 227 | |||
AK137000.1 | - | 3 | 299 | |||
AK145021.1 | - | 3 | 209 | |||
AK145316.1 | - | 3 | 183 | |||
AK161029.1 | - | 3 | 283 | |||
AK164138.1 | - | 3 | 241 | |||
AK167457.1 | - | 3 | 198 | |||
AK208560.1 | - | 2 | 132 | |||
BC020416.1 | - | 2 | 109 | |||
BC085276.1 | - | 3 | 307 | 30443236 | ||
L76158.1 | - | 3 | 205 | |||
X95351.1 | - | 3 | 212 |