Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199068.2 | NP_951031.2 | 1 | 455 | ||
GenBank mRNA | AK144418.1 | - | 1 | 348 | ||
AK147375.1 | - | 1 | 453 | |||
BB646926.1 | - | 1 | 455 | |||
BC060238.1 | - | 1 | 435 | |||
L26507.1 | - | 1 | 431 | |||
U95016.1 | - | 1 | 431 |