Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010712.2 | NP_034842.2 | 3 | 470 | ||
GenBank mRNA | AB221565.1 | - | 2 | 251 | ||
AF135415.1 | - | 2 | 255 | |||
BC049834.1 | - | 3 | 470 | 6493203 | ||
U89343.1 | - | 2 | 233 |