Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141813.1 | - | 3 | 386 | ||
AK164484.1 | - | 3 | 387 | |||
AK170166.1 | - | 2 | 161 | |||
AK172083.1 | - | 2 | 145 | |||
BC063768.1 | - | 3 | 368 | |||
BY230595.1 | - | 2 | 188 | |||
CJ066818.1 | - | 3 | 403 | |||
CV563459.1 | - | 2 | 145 | |||
U14166.1 | - | 3 | 409 |