Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010722.5 | NP_034852.2 | 3, 4 | 584 | ||
GenBank mRNA | BC042430.1 | - | 3, 4 | 589 | 3674002 | |
BC051985.1 | - | 3, 4 | 560 | 5695459 | ||
X54098.1 | - | 3, 4 | 543 |