Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145805.2 | NP_665804.1 | 6 | 1314 | ||
XM_017022505.1 | XP_016877994.1 | 6 | 1435 | |||
GenBank mRNA | AB593133.1 | - | 6 | 2836 | ||
AK001022.1 | - | 6 | 1233 | |||
BC011967.1 | - | 6 | 1220 | 3866562 | ||
BC012136.1 | - | 6 | 1232 | 4558235 |