Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281731.1 | NP_001268660.1 | 3 | 727 | ||
NM_147190.3 | NP_671723.1 | 2 | 526 | |||
NR_104035.1 | 2 | 526 | ||||
XM_005269220.2 | XP_005269277.1 | 2 | 362 | |||
XM_006719687.3 | XP_006719750.1 | 2 | 362 | |||
XM_011538975.2 | XP_011537277.1 | 2 | 362 | |||
XM_017020202.1 | XP_016875691.1 | 2 | 286 | |||
XM_017020203.1 | XP_016875692.1 | 3 | 650 | |||
XM_017020204.1 | XP_016875693.1 | 2 | 362 | |||
XM_017020205.1 | XP_016875694.1 | 2 | 215 | |||
GenBank mRNA | AK026014.1 | - | 2 | 332 | ||
AK058033.1 | - | 1 | 171 | |||
AK294521.1 | - | 2 | 300 | |||
AK300937.1 | - | 3 | 502 | |||
BC032565.1 | - | 2 | 343 | 5580489 | ||
BC033558.1 | - | 2 | 313 |