Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 6 | 1096 | ||
NM_001172767.2 | NP_001166238.1 | 6 | 810 | |||
NM_014491.3 | NP_055306.1 | 6 | 1099 | |||
NM_148898.3 | NP_683696.2 | 7 | 1174 | |||
NM_148899.3 | NP_683697.2 | 5 | 735 | |||
NM_148900.3 | NP_683698.2 | 7 | 1150 | |||
NR_033766.1 | 10 | 1312 | ||||
NR_033767.1 | 7 | 1157 | ||||
XM_017012801.1 | XP_016868290.1 | 9 | 1780 | |||
GenBank mRNA | AF337817.1 | - | 6 | 875 | ||
AF467253.1 | - | 5 | 737 | |||
AF467257.1 | - | 6 | 777 | |||
AF467258.1 | - | 5 | 722 | |||
AF467259.1 | - | 6 | 797 | |||
AK131266.1 | - | 10 | 1296 | |||
AK296957.1 | - | 7 | 1030 | |||
AY144615.1 | - | 7 | 1169 | |||
BC126104.1 | - | 7 | 831 | 8991820 | ||
BC143866.1 | - | 7 | 831 | 9052382 | ||
BC143867.1 | - | 6 | 770 | 9052383 | ||
CR749236.1 | - | 12 | 1325 | |||
DQ778626.1 | - | 3 | 449 | |||
KJ534839.1 | - | 6 | 800 | |||
KJ535079.1 | - | 5 | 938 | |||
KJ535086.1 | - | 5 | 724 |