Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288586.1 | NP_001275515.1 | 4, 5 | 499 | ||
NM_010786.4 | NP_034916.1 | 5, 6 | 584 | |||
GenBank mRNA | AK004719.1 | - | 5, 6 | 543 | ||
AK004881.1 | - | 5, 6 | 511 | |||
AK088638.1 | - | 5, 6 | 514 | |||
AK150530.1 | - | 4, 5 | 372 | |||
AK152685.1 | - | 4, 5 | 372 | |||
AK160069.1 | - | 4, 5 | 375 | |||
AK168690.1 | - | 5, 6 | 584 | |||
BC050902.1 | - | 5, 6 | 500 | 6415937 | ||
BC092270.1 | - | 5, 6 | 537 | 4018419 | ||
CN703560.1 | - | 4, 5 | 441 | |||
U47934.1 | - | 3, 4 | 290 | |||
X58876.1 | - | 5, 6 | 491 |