Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 5 | 476 | ||
GenBank mRNA | AK010498.1 | - | 5 | 443 | ||
AK011932.1 | - | 5 | 476 | |||
AK129076.1 | - | 5 | 429 | |||
AK137000.1 | - | 5 | 501 | |||
AK145021.1 | - | 5 | 411 | |||
AK145316.1 | - | 5 | 385 | |||
AK161029.1 | - | 5 | 485 | |||
AK164138.1 | - | 5 | 443 | |||
AK167457.1 | - | 5 | 400 | |||
BC085276.1 | - | 5 | 509 | 30443236 | ||
L76158.1 | - | 5 | 407 | |||
X95351.1 | - | 5 | 414 |