Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010791.3 | NP_034921.1 | 1 | 803 | ||
GenBank mRNA | AK052370.1 | - | 1 | 813 | ||
AK133672.1 | - | 1 | 174 | |||
BC011082.1 | - | 1 | 803 | 3984366 | ||
Z15103.1 | - | 1 | 758 |