Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010813.3 | NP_034943.3 | 5 | 1237 | ||
GenBank mRNA | AK139743.1 | - | 4 | 259 | ||
AK147933.1 | - | 5 | 1221 | |||
BC054534.1 | - | 5 | 1163 | 5721249 | ||
U77356.1 | - | 5 | 1237 | |||
Y07609.1 | - | 5 | 1183 |